Order Entry
Norway
ContactUsLinkComponent
Anti-cGAS (phospho Ser291) Rabbit Polyclonal Antibody
Anti-cGAS (phospho Ser291) Rabbit Polyclonal Antibody
Catalog # ANTIA308163-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-cGAS (phospho Ser291) Rabbit Polyclonal Antibody
Catalog # ANTIA308163-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C6orf150
  • Antigen symbol:
    cGAS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8C6L5
  • Antigen synonyms:
    CGAS_HUMAN|cGAMP synthase|Chromosome 6 open reading frame 150|cGAS|Mab 21 domain containing 1|Cyclic GMP-AMP synthase|cGMP Synthase|Hypothetical protein LOC115004|h-cGAS
  • Modification:
    phospho Ser291
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Molecular weight:
    59 kDa
  • Sequence:
    PGSPA
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around S291 of mouse CGAS (NP_775562.2)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to cGAS (phospho Ser291) for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: cGAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse