Order Entry
Norway
ContactUsLinkComponent
 
Anti-GAD65 Rabbit Polyclonal Antibody
  ANTIA12729-100
New Product
undefined
Anti-GAD65 Rabbit Polyclonal Antibody
  ANTIA12729-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    65 kDa glutamic acid decarboxylase
  • Antigen symbol:
    GAD65
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q05329
  • Antigen synonyms:
    GAD 65|GAD65|GAD-65|Gad2|DCE 2|GAD 2|GAD-2|DCE2|DCE2_HUMAN
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    60 kDa
  • Sequence:
    PLQCSALLVREEGLMQNCNQMHASYLFQQDKHYDLSYDTGDKALQCGRHVDVFKLWLMWRAKGTTGFEAHVDKCLELAEYLYNIIKNREGYEMVFDGKPQHTNVCFWYIPPSLRTLEDNEERMSRLSKVAPVIKARMMEYGTTMVSYQPLGDKVNFFRMVISNPAATHQDIDFLIEEIERLGQDL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 401-585 of human GAD65/GAD2 (NP_001127838.1)
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification.
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to GAD65 for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:1000, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: GAD65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat