Anti-ROR alpha/RORA Rabbit Polyclonal Antibody
ANTIA12052-100
New Product
- Antibody type:Primary
- Antigen symbol:ROR alpha/RORA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P35398
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
- Molecular weight:70 kDa
- Sequence:QPGEAEPLTPTYNISANGLTELHDDLSNYIDGHTPEGSKADSAVSSFYLDIQPSPDQSGLDINGIKPEPICDYTPASGFFP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 175 - 255 of human RORA (NP_599023.1)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to ROR alpha / RORA for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: ROR alpha/RORA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat