Order Entry
Norway
ContactUsLinkComponent
 
Anti-CFTR Mouse Recombinant Antibody [clone: rCFTR/7175]
  ANTIA316166-100
New Product
Anti-CFTR Mouse Recombinant Antibody [clone: rCFTR/7175]
  ANTIA316166-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ABC 35
  • Antigen symbol:
    CFTR
  • Clonality:
    Recombinant
  • Clone:
    rCFTR/7175
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13569
  • Antigen synonyms:
    ABC35|ABCC7|ATP-binding cassette sub-family C member 7|cAMP-dependent chloride channel|cAMP dependent chloride channel|ATP binding cassette transporter sub family C member 7|ABCC 7|ATP binding cassette sub family C member 7|ATP Binding Cassette Superfamily C Member 7
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    165 - 170 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 550-850) of human CFTR protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rCFTR/7175] antibody to CFTR for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: CFTR
Clonality: Recombinant
Clone: rCFTR/7175
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human