Anti-ROR gamma Mouse Monoclonal Antibody [clone: RORC/2941]
Catalog # ANTIA249911-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:IMD42
- Antigen symbol:ROR gamma
- Clonality:Monoclonal
- Clone:RORC/2941
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P51449
- Antigen synonyms:Nuclear receptor subfamily 1 group F member 3|MGC129539|RAR related orphan nuclear receptor variant 2|NR1F3|Nuclear receptor ROR gamma|RAR related orphan receptor C|Nuclear receptor RZR-gamma|Nuclear receptor ROR-gamma|Nuclear receptor RZR gamma
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:63 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human RORC protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [RORC/2941] antibody to ROR gamma for flow cytometry and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ROR gamma
Clonality: Monoclonal
Clone: RORC/2941
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human