Anti-MMP3 Rabbit Recombinant Antibody [clone: MMP3/1994R]
Catalog # ANTIA249372-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CHDS6
- Antigen symbol:MMP3
- Clonality:Recombinant
- Clone:MMP3/1994R
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08254
- Antigen synonyms:MMP-3|MMP 3|MMP3_HUMAN|Matrix metalloproteinase-3|MMP3|MGC126103|MGC126102|MGC126104|Matrix metalloproteinase 3
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 317-327, of human MMP3 protein. The exact sequence is proprietary.
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [MMP3/1994R] antibody to MMP3 for ELISA, flow cytometry and WB with samples derived from human.
- Validated applications: ELISA, Flow cytometry, WB
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml
Type: Primary
Antigen: MMP3
Clonality: Recombinant
Clone: MMP3/1994R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human