Anti-MMP3 Mouse Recombinant Antibody [clone: rMMP3/1730]
ANTIA249369-100
New Product
- Antibody type:Primary
- Antigen name:CHDS6
- Antigen symbol:MMP3
- Clonality:Recombinant
- Clone:rMMP3/1730
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08254
- Antigen synonyms:MMP-3|MMP 3|MMP3_HUMAN|Matrix metalloproteinase-3|MMP3|MGC126103|MGC126102|MGC126104|Matrix metalloproteinase 3
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human mMP3 protein
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rMMP3/1730] antibody to MMP3 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MMP3
Clonality: Recombinant
Clone: rMMP3/1730
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human