Order Entry
Norway
ContactUsLinkComponent
 
Mouse PFKM ELISA Kit
  ANTIA79846-96
New Product
 :  
undefined
Mouse PFKM ELISA Kit
  ANTIA79846-96
New Product
 :  

 

  • Fp:
    96 Tests
  • Analysens varighet:
    Multiple steps
  • Analysetype:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Mål:
    PFKM
  • Beskrivelse:
    Mouse PFKM ELISA kit
  • Type prøve:
    Serum, plasma, tissue homogenates and other biological fluids
  • Kryssreaktivitet:
    Mouse PFKM ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse PFKM. No significant cross-reactivity or interference between PFKM and analogues was observed.
  • Påvisningsmetode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analyseprinsipp:
    Quantitative
  • Holdbarhet:
    Store for 6 months at 4 °C
  • Påvisningsspekter:
    78,125 - 5000 pg/ml
  • Lagringstemp:
    4 °C
  • Prøvevolum:
    100 μl
  • Sensitivitet:
    46,875 pg/ml
  • Regulatorisk status:
    RUO

 

 

Mouse PFKM ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PFKM in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse PFKM ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PFKM in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PFKM has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PFKM present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PFKM Antibody, which binds the captured PFKM present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PFKM captured in each well. The concentration of PFKM can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.