Order Entry
Norway
ContactUsLinkComponent
 
Human PFKFB3 ELISA Kit
  ANTIA247428-96
New Product
 :  
undefined
Human PFKFB3 ELISA Kit
  ANTIA247428-96
New Product
 :  

 

  • Fp:
    96 Tests
  • Analysens varighet:
    Multiple steps
  • Analysetype:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Mål:
    PFKFB3
  • Beskrivelse:
    Human PFKFB3 ELISA kit
  • Type prøve:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Kryssreaktivitet:
    Human PFKFB3 ELISA Kit exhibits high specificity and excellent specificity for the detection of human PFKFB3. No significant cross-reactivity or interference between PFKFB3 and analogues was observed.
  • Påvisningsmetode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Holdbarhet:
    Store for 6 months at 4 °C
  • Påvisningsspekter:
    0,156 - 10 ng/ml
  • Lagringstemp:
    4 °C
  • Prøvevolum:
    100 μl
  • Sensitivitet:
    0,094 ng/ml
  • Regulatorisk status:
    RUO

 

 

Human PFKFB3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PFKFB3 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human PFKFB3 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PFKFB3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PFKFB3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PFKFB3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PFKFB3 Antibody, which binds the captured PFKFB3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PFKFB3 captured in each well. The concentration of PFKFB3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.