Specifications
- Fp:96 Tests
- Analysens varighet:One step
- Analysetype:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Mål:SYNRG
- Beskrivelse:Human SYNRG ELISA kit
- Type prøve:Serum, plasma or other biological fluids
- Påvisningsmetode:Colorimetric
- Time to Results:1 h 30 min
- Holdbarhet:Store for 6 months at 4 °C
- Påvisningsspekter:6 - 96 ng/ml
- Lagringstemp:4 °C
- Prøvevolum:40 μl
- Sensitivitet:0,27 ng/ml
- Regulatorisk status:RUO
Specifications
About this item
Human SYNRG ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SYNRG in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 6 to 96 ng/ml
- Sensitivity: 0,27 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human SYNRG ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SYNRG in An antibody specific for SYNRG has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SYNRG present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SYNRG antibody, which also binds the SYNRG present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SYNRG antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SYNRG antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of SYNRG captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SYNRG in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.