Human Factor V ELISA Kit
Product Details & Documents
Human Factor V ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Factor V in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 15,625 - 1000 pg/ml
- Sensitivity: 9,375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human Factor V ELISA kit (A79313) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Factor V in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Factor V has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Factor V present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Factor V Antibody, which binds the captured Factor V present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Factor V captured in each well. The concentration of Factor V can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- Pk:96 Tests
- Description:Human factor V ELISA kit
- Primary antibody reactivity:Human
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:100 μl
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Human Factor V ELISA kit exhibits high specificity and excellent specificity for the detection of human Factor V. No significant cross-reactivity or interference between Factor V and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:ANTIA79313-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:9,375 pg/ml
- Format:Pre-coated
- Target protein:Factor V
- Regulatory status:RUO



