Order Entry
Norway
Orders LinkContactUsLinkComponent
Human Complement C5b ELISA Kit

Human Complement C5b ELISA Kit

Human complement C5b ELISA kit, 96 Tests
Catalog # ANTIA77781-96
Supplier:   Antibodies.com
Human Complement C5b ELISA Kit
Catalog # ANTIA77781-96
Supplier:  Antibodies.com
CAS Number:  
Human Complement C5b ELISA Kit
Human Complement C5b ELISA Kit

Human complement C5b ELISA kit, 96 Tests

CAS Number:  
Price
kr7,685.00
Log in for availability
1 * 1 Each
Product Details & Documents

Human Complement C5b ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement C5b in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,781 to 50 ng/ml
  • Sensitivity: 0,469 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Complement C5b ELISA kit (A77781) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement C5b in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C5b has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C5b present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C5b Antibody, which binds the captured Complement C5b present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C5b captured in each well. The concentration of Complement C5b can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Human complement C5b ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    0,781 - 50 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Human Complement C5b ELISA kit exhibits high specificity and excellent specificity for the detection of human Complement C5b. No significant cross-reactivity or interference between Complement C5b and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA77781-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,469 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Complement C5b
  • Regulatory status:
    RUO

Specifications