Order Entry
Norway
ContactUsLinkComponent
Halofuginone hydrobromide
Halofuginone hydrobromide
Catalog # EHERC14059280
Supplier:  EHRENSTORFER
New Product
CAS Number:  
Halofuginone hydrobromide
Catalog # EHERC14059280
Supplier:  EHRENSTORFER
New Product
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :495.59 g/mol
CAS Number:64924-67-0
Formula:C₁₆H₁₇BrClN₃O₃·HBr
MDL Number:MFCD00946455