Order Entry
Norway
ContactUsLinkComponent
Human/Mouse Recombinant TGF-beta 3
Human/Mouse Recombinant TGF-beta 3
Certificates
undefined
Human/Mouse Recombinant TGF-beta 3

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
    Mouse
  • Protein/peptide name:
    TGF-beta 3
  • Purity:
    ≥95%

Specifications