Order Entry
Norway
ContactUsLinkComponent
Anaerobic systems, workstation, Whitley A35
Anaerobic systems, workstation, Whitley A35
Certificates
undefined
Anaerobic systems, workstation, Whitley A35
Specifications
  • Catalog No:
  • 710-0842
  • 710-0843
  • Capacity:
  • Working: 560 plates;
    Incubation: 620 plates;
    Port/air lock: 40 plates
  • Working: 560 plates;
    Incubation: 620 plates;
    Port/air lock: 40 plates
  • Cycle time:
  • 60 s (airlock)
  • 60 s (airlock)
  • Gas type:
  • ANO2 / N2
  • ANO2 / N2