Test Strips, Zinc
About this item
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of Zinc in solutions. They come with all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 2 minutes.
8 Options Available Below
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
- Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips allow the quick and easy determination of Zinc in solutions. They come with all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 2 minutes.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 662-0239
- 662-1867
- MANA91239
- MANA91223
- MANA91224
- MANA91240
- MANA912902
- MANA91221
- Method:
- Total hardness - green → red
- Total hardness - green → red
- Total hardness - green → red
- Total hardness - green → red
- Total hardness - green → red
- Total hardness - green → red
- Total hardness - green → red
- Total hardness - green → red
- Parameters:
- All reagents included
- All reagents included
- All reagents included
- All reagents included
- All reagents included
- All reagents included
- All reagents included
- All reagents included
- Compliance:
- No hazardous material
- No hazardous material
- No hazardous material
- No hazardous material
- No hazardous material
- No hazardous material
- No hazardous material
- No hazardous material
- Contents:
- 100 test strips in a tube
- 100 test strips in a tube
- 100 test strips in a tube
- 1000 individually sealed test strips in a box
- 1000 individually sealed test strips in a box
- 1000 individually sealed test strips in a box
- 50 sets of 3 individually sealed test strips each
- 5000 test strips in a box without color chart
- Instrument:
- No
- No
- No
- No
- No
- No
- No
- No
- Platform:
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Semi-quantitative test strips
- Shelf life (months):
- 24 (from production)
- 24 (from production)
- 24 (from production)
- 12 (from production)
- 12 (from production)
- 12 (from production)
- 12 (from production)
- 15 (from production)
- Weight:
- 40 g
- 40,6 g
- 40,2 g
- 596,7 g
- 598,2 g
- 613 g
- 205,99 g
- 796,6 g
Specifications
Product Family Options
Product Information
- RangeDimensionsTests per kitPkAvailabilityPrice
- 662-0239AQUADUR® 4-21AQUADUR® 4-21< 54 • >72 • >126 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >7 • >14 • >21 - °e40×40×126 mm (packaging)100 Tests100Log in for availability1 * 100 Itemskr571.00Specifications:
More
Specifications
Cat. No.662-0239MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tubeInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)24 (from production)Weight40 g - 662-1867AQUADUR® 5-25AQUADUR® 5-25< 54 • >90 • >180 • >270 • >360 • >450 - ppm - CaCO₃; <3 • >5 • >10 • >15 • >20 • >25 - °e40×40×123 mm (packaging)100 Tests100Log in for availability1 * 100 Itemskr571.00Specifications:
More
Specifications
Cat. No.662-1867MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tubeInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)24 (from production)Weight40,6 g - AQUADUR® 4-14< 54 • >72 • >151,2 • >252 - ppm - CaCO₃40×40×120 mm (packaging)100 Tests1Log in for availability1 * 1 Itemskr871.00Specifications:
More
Specifications
Cat. No.MANA91239MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents100 test strips in a tubeInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)24 (from production)Weight40,2 g - AQUADUR® 5-25< 54 • >90 • >180 • >270 • >360 • >450 - ppm - CaCO₃; <3 • >5 • >10 • >15 • >20 • >25 - °e210×215×135 mm (packaging)1000 Tests1.000Log in for availability1 * 1.000 Itemskr5,140.00Specifications:
More
Specifications
Cat. No.MANA91223MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents1000 individually sealed test strips in a boxInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)12 (from production)Weight596,7 g - AQUADUR® 4-21< 54 • >72 • >126 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >7 • >14 • >21 - °e210×210×135 mm (packaging)1000 Tests1.000Log in for availability1 * 1.000 Itemskr5,140.00Specifications:
More
Specifications
Cat. No.MANA91224MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents1000 individually sealed test strips in a boxInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)12 (from production)Weight598,2 g - AQUADUR® 4-21< 54 • >72 • >151,2 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >8,4 • >14 • >21 - °e223×215×142 mm (packaging)1000 Tests1.000Log in for availability1 * 1.000 Itemskr5,140.00Specifications:
More
Specifications
Cat. No.MANA91240MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents1000 individually sealed test strips in a boxInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)12 (from production)Weight613 g - MANA912902AQUADUR® 5-25 test setsAQUADUR® 5-25 test sets< 54 • >90 • >180 • >270 • >360 • >450 - ppm - CaCO₃93×50×125 mm (packaging)150 Tests1 KITLog in for availability1 * 1 Eachkr1,800.00Specifications:
More
Specifications
Cat. No.MANA912902MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents50 sets of 3 individually sealed test strips eachInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)12 (from production)Weight205,99 g - AQUADUR® 5-25< 54 • >90 • >180 • >270 • >360 • >450 - ppm - CaCO₃270×130×60 mm (packaging)5000 Tests5.000Log in for availability1 * 5.000 Itemskr9,840.00Specifications:
More
Specifications
Cat. No.MANA91221MethodTotal hardness - green → redParametersAll reagents includedComplianceNo hazardous materialContents5000 test strips in a box without color chartInstrumentNoPlatformSemi-quantitative test stripsShelf life (months)15 (from production)Weight796,6 g