Eosin Y (yellowish), Certifiable Grade
Catalog # 0109-100G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Bottle, amber
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Dye Content:>88%
- Lambda Max:515 – 518 nm
Specification Test Results
About this item
:
Merck Index:13,03634
Melting Pt:295…296 °C
:691.86 g/mol
EINECS:241-409-6
:GHS07
Storage Temperature:Ambient
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040