Eosin Y, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:17372-87-1
:
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040
Merck Index:13,03634
Density:1,018 g/cm³
:M-Clarity™ quality level = MQ200
Flash Pt:191,6 °C
:691.86 g/mol
EINECS:241-409-6
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Solubility (Colour) 1 mg/ml H₂O:Orange to Very Dark Orange and Red to Very Dark Red and Orange-Red and Red-Orange
- Solubility (Turbidity):Clear
- Dye Content:≥85%
- pH:6.55 (29 °C, 84 °F)