Order Entry
Norway
ContactUsLinkComponent
Sodium nitrate 99.99%, HighPurS-CERT
Oxidizing
Irritant
Sodium nitrate 99.99%, HighPurS-CERT
Catalog # ACSDPSNANAC.50G
Supplier:  CPACHEM
New Product
CAS Number:  
Oxidizing
Irritant
Sodium nitrate 99.99%, HighPurS-CERT
Catalog # ACSDPSNANAC.50G
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :84.99 g/mol
Boiling Pt:600...700 °C
Melting Pt:312 °C
Density:2,26 g/cm³
EINECS:231-554-3
UN:1498
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:7631-99-4
Formula:NaNO₃
MDL Number:MFCD00011119