BrainPhys™ Imaging Optimised Medium
Catalog # STMC05796
Leverandør: STEMCELL Technologies
CAS-nummer:
Spesifikasjoner
- Pk:500 mL
- Description:BrainPhys™ Imaging optimized medium
Spesifikasjoner
Om dette produktet
Serum-free neurophysiological basal medium for improved neuronal live imaging and function.
BrainPhys™ Imaging optimised Medium is a serum-free neuronal basal medium that is optimised for imaging applications. Applications of BrainPhys™ Imaging optimised Medium include live fluorescent imaging (calcium imaging and optogenetics) and neuronal culture. In addition to the removal of phenol red, the formulation has been modified to reduce background fluorescence and increase stability upon repeated exposure to light.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').