Specifications
- Pk:1 mL
- Description:StemSpan™ CC110
Specifications
About this item
Serum-free culture supplement for expansion of human hematopoietic cells.
StemSpan™ CC110 contains a combination of early-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate. When added to serum-free medium, StemSpan™ CC110 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow. StemSpan™ CC110 stimulates similar expansion of CD34+ cells compared to StemSpan™ CC100, but with higher purity and is meant for use in short-term cultures to activate stem cell and immature progenitor cycling, without necessarily promoting the proliferation and differentiation of later progenitors.
We recommend using StemSpan™ CC110 in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF