Order Entry
Norway
ContactUsLinkComponent
MesenCult™ MSC Stimulatory Supplement (Human)
MesenCult™ MSC Stimulatory Supplement (Human)
Catalog # STMC05402
CAS Number:  
undefined
MesenCult™ MSC Stimulatory Supplement (Human)
Catalog # STMC05402
CAS Number:  
Specifications
  • Fp:
    50 mL
  • Beskrivelse:
    MesenCult™ MSC stimulatory supplement (human)
Specifications
About this item
Supplement for detection of CFU-F and expansion of human mesenchymal stem cells.
MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is a standardised, serum-containing supplement for the culture of human mesenchymal stem cells (MSCs). MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human) is optimised for the expansion of human mesenchymal stem cells in vitro as well as their enumeration using the colony-forming unit - fibroblast (CFU-F) assay when combined with MesenCult™ MSC Basal Medium (Human). Components are pre-screened to minimise lot-to-lot variability. This supplement is also a component of MesenCult™ Proliferation Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').