Anti-FBXW12 Rabbit Polyclonal Antibody
Catalog # PRSI55-114
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:F-box/WD repeat-containing protein 12
- Antigen symbol:FBXW12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino Acid Range = 291-320
- Form:Liquid
- Gene ID:Q6X9E4
- Antigen synonyms:FBXW12
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:53 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This FBXW12 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 291-320 amino acids from the C-terminal region of human FBXW12.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
Members of the F-box protein family, such as FBXW12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603034), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM].
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: FBXW12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human