Anti-UNC93B1 Rabbit Polyclonal Antibody
PRSI33-023
New Product
- Antibody type:Primary
- Antigen name:unc-93 homolog B1 (C. elegans)
- Antigen symbol:UNC93B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q9H1C4
- Antigen synonyms:UNC93B1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at –20 °C. Avoid freez-thaw cycles.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:A recombinant protein fragment from the C-terminal portion of UNC93B was used as the immunogen for this antibody.
- Purification:Protein A Affinity Chromatography
- Size:100 µg
- Pk:100 µG
UNC93B is part of the innate immune response via its direct interaction with the intracellular toll-like receptors TLR3, 7, and 9. UNC93B aids in the trafficing of these TLRs from the ER to the endolysosomes, where they bind ligands and initiate immune response. In the absence of UNC93B, these TLRs are unable to respond to foreign stimuli.
Western blot: 3-6 µg/ml Provided assay concentrations are sµggestions only, antibody titration may be required for optimal results.
Type: Primary
Antigen: UNC93B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog