Anti-LPCAT4 Rabbit Polyclonal Antibody
PRSI15-481
New Product
- Antibody type:Primary
- Antigen name:Lysophosphatidylcholine acyltransferase 4
- Antigen symbol:LPCAT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:254531
- Antigen synonyms:LPEAT2|lysophospholipid acyltransferase LPCAT4|AGPAT7|1-acylglycerol-3-phosphate O-acyltransferase 7 (lysophosphatidic acid acyltransferase|plasmalogen synthase|acyltransferase-like 3|1-acylglycerophosphocholine O-acyltransferase|1-AGPAT 7|eta)|AYTL3|1-alkenylglycerophosphoethanolamine O-acyltransferase|1-alkylglycerophosphocholine O-acetyltransferase|PLSC domain containing protein|LPAAT-eta|lysophosphatidylethanolamine acyltransferase 2|1-acylglycerophosphoserine O-acyltransferase|acyl-CoA:lysophosphatidylethanolamine acyltransferase 2|1-AGP acyltransferase 7
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 345-524 of human LPCAT4 (NP_705841.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: LPCAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse