Anti-LPCAT2 Rabbit Polyclonal Antibody
PRSI19-475
New Product
- Antibody type:Primary
- Antigen name:Lysophosphatidylcholine acyltransferase 2
- Antigen symbol:LPCAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:54947
- Antigen synonyms:AYTL1|1-AGP acyltransferase 11|lysophosphatidylcholine acyltransferase 2|LysoPAFAT|AGPAT11|acyl-CoA:lysophosphatidylcholine acyltransferase 2|LPCAT-2|acyltransferase-like 1|1-AGPAT 11|acetyl-CoA:lyso-PAF acetyltransferase|LPC acyltransferase 2|1-acylglycerophosphocholine O-acyltransferase|1-acylglycerol-3-phosphate O-acyltransferase 11|lysoPC acyltransferase 2|1-alkylglycerophosphocholine O-acetyltransferase|lyso-PAF acetyltransferase|lysophosphatidic acid acyltransferase alpha|lyso-platelet-activating factor (PAF) acetyltransferase|acetyl-CoA:lyso-platelet-activating factor acetyltransferase|LPAAT-alpha
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 160-544 of human LPCAT2 (NP_060309.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: LPCAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human