Order Entry
Norway
ContactUsLinkComponent
Human Recombinant PDI-associated 1 (from E. coli)
Human Recombinant PDI-associated 1 (from E. coli)
Catalog # PRSI92-109
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant PDI-associated 1 (from E. coli)
Catalog # PRSI92-109
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    ERBA2L|PDIA1|Protein Disulfide-Isomerase|p55|Prolyl 4-Hydroxylase Subunit Beta|PDI|Cellular Thyroid Hormone-Binding Protein|PO4DB|P4HB
  • Protein/peptide name:
    PDI-associated 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    55,9 kDa
  • Sequence:
    Asp18-Lys505
  • Formulation:
    Supplied as a 0,2 µm filtered solution of PBS, pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Protein Disulfide-Isomerase (P4HB) is an endoplasmic reticulum lumen protein that belongs to the protein disulfide isomerase family. P4HB contains two thioredoxin domains and catalyzes the formation, breakage, and rearrangement of -S-S- bonds in proteins. P4HB is involved in hydroxylation of prolyl residues in preprocollagen. P4HB has the ability to act as a chaperone that inhibits aggregation of misfolded proteins in a concentration-dependent manner. P4HB plays a role in both the influx and efflux of S-nitrosothiol-bound nitric oxide.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays.

Caution: For research use only.