Order Entry
Norway
ContactUsLinkComponent
Anti-Tlr2 Mouse Monoclonal Antibody [clone: mT2.7] (FITC (Fluorescein Isothiocyanate))
Anti-Tlr2 Mouse Monoclonal Antibody [clone: mT2.7] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-951
Supplier:  ProSci Inc.
Anti-Tlr2 Mouse Monoclonal Antibody [clone: mT2.7] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-951
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    toll-like receptor 2
  • Antigen symbol:
    Tlr2
  • Clonality:
    Monoclonal
  • Clone:
    mT2.7
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    Q9QUN7
  • Antigen synonyms:
    Tlr2
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

The mT2.7 monoclonal antibody specifically reacts with mouse CD282, the Toll-like receptor 2 (TLR2). It is a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. CD282 is expressed on monocytes, macrophages, dendritic cells, and the RAW264.7 cell line and is involved distinguishing bacterial lipoproteins.

Type: Primary
Antigen: Tlr2
Clonality: Monoclonal
Clone: mT2.7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse