Order Entry
Norway
ContactUsLinkComponent
Anti-SDC1 Mouse Monoclonal Antibody [clone: DL-101] (PE (Phycoerythrin))
Anti-SDC1 Mouse Monoclonal Antibody [clone: DL-101] (PE (Phycoerythrin))
  PRSI76-934
Anti-SDC1 Mouse Monoclonal Antibody [clone: DL-101] (PE (Phycoerythrin))
  PRSI76-934

 

  • Antibody type:
    Primary
  • Antigen name:
    syndecan 1
  • Antigen symbol:
    SDC1
  • Clonality:
    Monoclonal
  • Clone:
    DL-101
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P18827
  • Antigen synonyms:
    SDC1
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    25 Tests

 

 

The DL-101 monoclonal antibody specifically reacts with human CD138, a 85-92 kDa type I heparan sulfate proteoglycan. CD138 is known as Syndecan-1 and is expressed on pre-B cells, immature B cells, plasma cells, and malignant plasma cells, to the exclusion of mature circulating B cells. Syndecan-1 is also expressed on vascular smooth muscle, endothelial, neural, and embryonic mesenchymal cells. It is involved in cellular proliferation, cellular migration, and acts as an extracellular matrix receptor. Among the hematopoietic elements, CD138 is a useful marker for plasma cells.

Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: DL-101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human