Anti-BAX069 Recombinant Antibody
Catalog # PRSI10-668
Leverandør: ProSci Inc.
Spesifikasjoner
- Antibody type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Isotype:IgG2b kappa
- Antigen synonyms:BT 5/9|SAND-26
- Modification:None
- Storage buffer:PBS buffer pH 7,5
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Mus musculus/DPP4/TP103/CD26 [Homo sapiens]
- Purification:>95%
- Pack type:Vial
- Pk:0,1 mg
Spesifikasjoner
Om dette produktet
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b-kappa
Reactivity: