Anti-WBSCR22 Rabbit Polyclonal Antibody
PRSI60-954
New Product
- Antistofftype:Primary
- Antigen-navn:Probable 18S rRNA (guanine-N(7))-methyltransferase
- Antigensymbol:WBSCR22
- Klonalitet:Polyclonal
- Konjugering:Unconjugated
- Vert:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:253 - 281 aa
- Skjema:Liquid
- Gen-ID:114049
- Modifikasjon:None
- Molekylvekt:32 kDa
- Lagringstemp:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konsentrasjon:0,4 mg/ml
- Frakttemp.:4 °C
- Immunogen:This WBSCR22 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 253 - 281 amino acids from the C-terminal region of human WBSCR22.
- Rensing:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Fp:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: WBSCR22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 253-281
Host: Rabbit
Isotype: Ig
Reactivity: Human