Anti-MLLT10 Rabbit Polyclonal Antibody
Catalog # PRSI60-305
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Protein AF-10
- Antigen symbol:MLLT10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Epitope:294 - 323 aa
- Form:Liquid
- Gene ID:8028
- Modification:None
- Molecular weight:113 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,35 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MLLT10 (AF10) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 294 - 323 amino acids from the Central region of human MLLT10 (AF10).
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: MLLT10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 294-323
Host: Rabbit
Isotype: Ig
Reactivity: Human