Anti-TSPAN1 Recombinant Antibody [Clone: 4/12]
Catalog # PRSI24-725
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:tetraspanin 1
- Antigen symbol:TSPAN1
- Clonality:Recombinant
- Clone:4/12
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:10103
- Antigen synonyms:Tetraspanin-1|Tetraspanin TM4-C|TSPAN1|TM4SF|Tspan-1|C4.8|Tetraspan NET-1|Net1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: tetraspanin 1
Clonality: Recombinant
Clone: 4/12
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human