Order Entry
Norway
ContactUsLinkComponent
 
Oil of cedar wood
Catalog # CHRO3D-078
Supplier:  CHROMA
CAS Number:  
Oil of cedar wood
Catalog # CHRO3D-078
Supplier:  CHROMA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Density:0.98 g/cm³ (20 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:8000-27-9
 :68990-83-0
MDL Number:MFCD00146327