Organic reference standard, Vinyl acetate 5,000 µg/ml in acetonitrile, Supelco®
Catalog # 40327.
Supplier: Merck
CAS Number:
Specifications
- Fp:1
- CAS-nr:108-05-4
- Beskrivelse:Vinyl acetate 5,000 µg/ml in acetonitrile
- Formel:C₄H₆O₂
- Beskrivelse:Vinyl acetate