Order Entry
Norway
ContactUsLinkComponent
 
Calcon® (mordant black 17), Supelco®
  1.04594.0050
 :  Merck
 :  
Calcon® (mordant black 17), Supelco®
  1.04594.0050
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

 

 

  • Solubility:
    20 g/l (20 °C)
  • Molar Mass:
    416.39 g/mol
  • Bulk Density:
    530 kg/m3
  • pH:
    9.4 (10 g/l, H2O, 25 °C)

 

 

 :
 :416.38 g/mol
 :M-Clarity™ quality level = MQ200
 :Ambient
 :219-810-2
 :2538-85-4
 :C₂₀H₁₃N₂NaO₅S
 :MFCD00004069