α-Methylstyrene
Catalog # EHERCA15143500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:118.18 g/mol
Boiling Pt:165.4 °C (1013 hPa)
Melting Pt:–23 °C
Density:0.9108 g/cm³ (20 °C)
Flash Pt:43 °C
Storage Temperature:Refrigerator
EINECS:202-705-0
UN:2303
ADR:3,III
:GHS02
:GHS07
:GHS09
CAS Number:98-83-9
Formula:C₉H₁₀
MDL Number:MFCD00008859