Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(+/-)-Anatoxin
(+/-)-Anatoxin ≥98% (by TLC)
(+/-)-Anatoxin ≥98% (by TLC)
ENZOBMLC1180001
:
ENZO LIFE SCIENCES
:
(+/-)-Anatoxin ≥98% (by TLC)
ENZOBMLC1180001
:
ENZO LIFE SCIENCES
:
Pk:
1 mg
Seq:
2070
Purity:
≥98% (TLC)
Appearance:
White solid
:
:
281.3 g/mol
:
Freezer
:
3462
:
6.1,I
:
GHS06
:
1219922-30-1
:
C₁₀H₁₅NO·C₄H₄O₄