Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-1,8-naphthalimide
4-Amino-1,8-naphthalimide ≥96% (by NMR)
4-Amino-1,8-naphthalimide ≥96% (by NMR)
4-Amino-1,8-naphthalimide ≥96% (by NMR)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
4-Amino-1,8-naphthalimide ≥96% (by NMR)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
1742-95-6
:
Formula:
C₁₂H₈N₂O₂
MDL Number:
MFCD00006921
:
212.21 g/mol