Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Cyano-7-nitroquinoxaline-2,3-dione
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
115066-14-3
:
Formula:
C₉H₄N₄O₄
MDL Number:
MFCD00069232
:
232.16 g/mol