
DPTA NONOate ≥98% (by NMR)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:146724-95-0
:
Formula:C₆H₁₇N₅O₂
:191.2 g/mol
:GHS07
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
Specification Test Results
- Purity (NMR):≥98%