Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gentamicin sulphate
Gentamicin sulphate
Gentamicin sulphate
Gentamicin sulphate
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Gentamicin sulphate
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
1405-41-0
:
MDL Number:
MFCD00270181
Merck Index:
13,04403
Melting Pt:
218…237 °C
EINECS:
215-778-9
:
GHS08
Storage Temperature:
Ambient