Cholesterol-3-sulphate sodium salt
Catalog # ENZOALX370009M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:One spot by TLC
- Identity:Determined by IR
Specification Test Results
About this item
:
:488.7 g/mol
Storage Temperature:Refrigerator
CAS Number:2864-50-8
Formula:C₂₇H₄₅O₄S·Na
MDL Number:MFCD00079091