Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rotenone
Rotenone ≥98% (by HPLC)
Rotenone ≥98% (by HPLC)
:
ENZO LIFE SCIENCES
Rotenone ≥98% (by HPLC)
:
ENZO LIFE SCIENCES
:
83-79-4
:
:
C₂₃H₂₂O₆
:
MFCD09025614
:
394.42 g/mol