Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Salubrinal
Salubrinal ≥98% (by HPLC)
Salubrinal ≥98% (by HPLC)
Salubrinal ≥98% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Salubrinal ≥98% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
405060-95-9
:
Formula:
C₂₁H₁₇Cl₃N₄OS
MDL Number:
MFCD00548612
:
479.81 g/mol