Clofibrate ≥98% (by GC)
Catalog # ENZOALX270256G001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity:≥98% (GC)
Specification Test Results
About this item
:
Merck Index:14,02377
ADR:9,III
:242.70 g/mol
UN:3082
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:637-07-0
Formula:C₁₂H₁₅ClO₃
MDL Number:MFCD00000615