Order Entry
Norway
ContactUsLinkComponent
BML-280 ≥98% (by TLC)
BML-280 ≥98% (by TLC)
Catalog # ENZOBMLST3710005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
BML-280 ≥98% (by TLC)
Catalog # ENZOBMLST3710005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :429.5 g/mol
Storage Temperature:Freezer
CAS Number:1158347-73-9
Formula:C₂₅H₂₇N₅O₂