Order Entry
Norway
ContactUsLinkComponent
FH535 ≥98% (by TLC)
FH535 ≥98% (by TLC)
Catalog # ENZOBMLWN1120005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
FH535 ≥98% (by TLC)
Catalog # ENZOBMLWN1120005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :361.2 g/mol
CAS Number:108409-83-2
Formula:C₁₃H₁₀Cl₂N₂O₄S
MDL Number:MFCD01212888