kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
(±)-Lisofylline
(±)-Lisofylline ≥98%
(±)-Lisofylline ≥98%
(±)-Lisofylline ≥98%
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
(±)-Lisofylline ≥98%
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
6493-06-7
:
Formula:
C₁₃H₂₀N₄O₃
MDL Number:
MFCD00867573
:
280.3 g/mol