Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
BML-278
BML-278 ≥98% (by TLC)
BML-278 ≥98% (by TLC)
BML-278 ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
BML-278 ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
15301-69-6
:
Formula:
C₂₄H₂₅NO₄
:
391.5 g/mol
Storage Temperature:
Ambient
Specification Test Results
Purity:
≥98% by TLC
Specification Test Results